Would you like to log in again?
Eisco LDPE Dropping Bottles, Pack of 12
Supplier: EISCO SCIENTIFIC LLC
CH0182EPK12 CH0182CPK12 CH0182APK12 CH0182DPK12 CH0182BPK12
470346-044EA 27.7 USD InStock
470346-044 470346-088 470344-682 470345-586 470343-740
Eisco LDPE Dropping Bottles, Pack of 12
Bottles Dropping and Dispensing Bottles
Dispenses precise uniform drops of liquid.
  • Made of translucent low density polyethylene plastic (LDPE)
  • Screw cap with a built in dropper nozzle allows for small, uniform drops of liquid
  • Attached cover for nozzle prevents spills
Nozzle cover teathered to cap prevents leakage and spills.
Packaging: Comes in packs of 12 bottles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty