Would you like to log in again?
Extra Long Crucible Tongs
Supplier: EISCO SCIENTIFIC LLC
CH0732A
470343-754EA 8.25 USD InStock
470343-754
Extra Long Crucible Tongs
Tongs
Designed for furnace use, this extra long 13" stainless steel tong with straight, serrated tips provides a sturdy and reliable grip on crucibles.
  • Crucible tongs with extra long handle
  • Straight with serrated tips for added grip strength
  • Made of high-quality rust- and corrosion-resistant stainless steel
Long and rigid design makes tongs excelled for use in a furnace or other similar equipment. Designed for use in professional laboratories, but is an ideal choice for science classrooms. Tongs measure 13" long, 4" wide when collapsed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty