Would you like to log in again?
Eisco Glass Thistle Tube Funnels
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0489C CH0489A CH0489B
470345-748EA 4.29 USD InStock
470345-748 470343-836 470344-226
Eisco Glass Thistle Tube Funnels
Funnels Filtration Funnels Filter Funnels
Stout-walled Thistle Tube Funnels for the delivery of liquid to an existing system or apparatus.
  • Made of high quality borosilicate 3.3 glass
  • Stout wall design for durability
  • Ideal for filling narrow items like burettes, barometer tubes, and manometers
  • Minimizes the risk of a reaction occurring too rapidly
Thistle Tube funnel for the insertion of liquid to an exact position in an existing system or apparatus. Reservoir is 38 mm in height with a 38 mm Ø and an Int.Ø of 25 mm. The Ext.Ø of the stem is 12 mm.
Eisco borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty