Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rectangular Double Burette Base

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0656A
470343-840EA 56.95 USD InStock
470343-840
Rectangular Double Burette Base
Support Stands
Rectangular double burette base with a 13×7" heavy-duty cast iron base in blue. The base offers stability and support for multiple burettes in laboratory settings.

  • Rectangular double burette base made of chemical-resistant cast iron with a blue coating
  • The 13" long by 7" wide base weighs 6.6 lbs., providing superb stability for experiments and setups
  • Base features a tapped hole for a 10×1.5 mm threaded rod (sold separately)
  • Best used with Eisco labs rod series: CH0657, CH0658, CH0659
  • Durable and sturdy construction of this set makes it an excellent choice for science classrooms

Rectangular retort base or double burette stands are used in every area of science, especially chemistry, for holding and manipulating an experimental apparatus. This retort stand is made of chemical-resistant cast iron, and features a tapped hole, suitable for rods with a 10×1.5 mm metric thread. Stand measures 13" long, 7" wide, and approximately 1" in height. Stand weighs 6.6 lbs., providing superb stability for basic experiments and setups. The durable and sturdy construction of this base makes it an excellent choice for science classrooms and for basic professional laboratory setups.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty