Would you like to log in again?
Single Burette Clamp
Supplier: EISCO SCIENTIFIC LLC
CH0256A
470343-856EA 10.7 USD InStock
470343-856
Single Burette Clamp
Clamps General Purpose Clamps
Single burette clamp made of casted material with a built-in boss head.
  • Casted, with built-in boss head
  • Approximately 3.75" tall, 4.5" wide and 2.75" long
  • To secure single burette
  • Durable, non slip
  • Basic essential for classroom and laboratories
Fits burette up to 25 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty