Would you like to log in again?
Eisco Micro Bunsen Burner with Flame Stabilizer
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CH0093D CH0093E
470343-868EA 44.45 USD InStock
470343-868 470345-826
Eisco Micro Bunsen Burner with Flame Stabilizer
Heaters Gas Burners
Adjustable orifice and flame stabilizing burner tip.
  • Equipped with a flame stabilizing burner tip and air ports with collar to adjust the flow of air
  • Gas flow is controlled by needle valve fitted above the base permitting easy flame adjustment
  • Suitable for use with LPG (liquid petroleum gas) / butane gas or mixed gas with a heat value of 800 to 1200 BTU
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Aluminum mixing tube is mounted on a circular, nickel plated die casted zinc alloy base
Nickel-plated die casted zinc alloy base, aluminum mixing tube, flame stabilizing burner tip and adjustable air ports. Air supply is controlled by a nickel plated brass sleeve covering ports at the lower end of the mixing tube, and gas flow is controlled by needle valve fitted above the base for easy flame adjustment. Burner measures 3.75" in height and base measures 3" in Ø.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty