Would you like to log in again?
Volumetric Pipettes, Class A
Supplier: EISCO SCIENTIFIC LLC
CH0559C CH0559G CH0559F CH0559I CH0559A CH0559E CH0559H CH0559D CH0559B
470343-826EA 6.99 USD InStock
470343-826 470312-906 470312-908 470344-590 470312-910 470343-886 470312-912 470345-068 470312-914
Volumetric Pipettes, Class A
Pipettes Graduated Pipettes
Reusable Class-A pipettes made from high quality borosilicate 3.3 glass.
  • Color coded for easy identification
  • Single graduation mark
  • Calibrated for delivery
  • Autoclavable
These Class-A volumetric pipettes are perfect for everyday laboratory use. They are designed for transferring liquid substances from one vessel to another with high accuracy.
Certifications: Calibrated according to DIN ISO 648 standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty