Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Tubings, Pack of 10

Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0747D CH0747C
470343-902EA 15.1 USD InStock
470343-902 470345-804
Eisco Glass Tubings, Pack of 10
Tubing Transfer Tubing
Glass tubings made of high-quality borosilicate glass ideal for a variety of lab applications.

  • With superior durability and chemical resistance
  • Can be bent and shaped to suit specific lab needs using a high heat source
  • Each piece of tubing is approximately 50 cm in length

Pack of 10 glass tubings made of high-quality borosilicate 3.3 glass with light walls. Ideal for the passage of various gases and liquids during different scientific experiments.

Eisco borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty