Would you like to log in again?
Eisco Amber Glass Dropping Bottles HDPE Stopper Head
Supplier: EISCO SCIENTIFIC LLC
CH0195C CH0195B CH0195A
470343-922EA 15.05 USD InStock
470343-922 470345-270 470344-518
Eisco Amber Glass Dropping Bottles HDPE Stopper Head
Bottles Dropping and Dispensing Bottles
Highly resistant to acids and alkalis.
  • Made of high quality, heavy-duty, borosilicate 3.3 glass
  • 3-piece pipette section consists of a glass dropper, HDPE stopper with dust-proof head, and rubber bulb
  • Amber color glass appropriate for holding light sensitive contents
  • Suitable for a variety of applications in the classroom or laboratory
Equipped with an eye dropper pipette for clean dispensing of stored contents. The amber color glass absorbs UV waves, making it useful for light sensitive contents. Excellent for chemistry applications, transferring and mixing indicators, stains or other aqueous solutions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty