Would you like to log in again?
Eisco Economy Ballistic Pendulum
Supplier: EISCO SCIENTIFIC LLC
BLPN14
470343-924EA 99.85 USD InStock
470343-924
Eisco Economy Ballistic Pendulum
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Accurate apparatus for demonstrating Newton's laws of motion.
  • Includes ballistic launcher, rigid arm pendulum, angle backdrop, steel ball, and release mechanism
  • Convenient angle backdrop makes measurements easier
  • White scale against black surface for maximum contrast
The ballistic pendulum contains everything needed to measure the momentum of a projected object - the ballistic launcher, rigid arm pendulum, angle backdrop, steel ball, and release mechanism are all included. This unit can be used to aid in the understanding of velocity and acceleration, conservation of momentum and energy, potential energy, and kinetic energy. Great for use with photogates (not included). Its heavy base, sturdy construction and high-constrast scales make it easy to use and transport.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty