Would you like to log in again?
Crucible Tongs with Straight Tips
Supplier: EISCO SCIENTIFIC LLC
CH0729B
470343-950EA 3.89 USD InStock
470343-950
Crucible Tongs with Straight Tips
Tongs
These metal tongs feature a bow design and straight serrated tips. They provide a secure grip for crucibles in laboratory settings.
  • General-purpose, metal crucible tongs
  • Straight, serrated tips and bow
  • Tongs measure 9.5" in length
  • Perfect for any classroom or laboratory
General-purpose metal crucible tongs with small bow. Straight, serrated tips. Useful for grasping hot crucibles, evaporating dishes, or small flasks and beakers. Tongs measure 9.5" in total length, and 3.25" wide when closed. Each tip is approximately 0.9" long, and bow can hold items with a minimum diameter of 1".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty