Would you like to log in again?
Eisco Support Rings with Bosshead Clamp, Steel
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0672B CH0672A CH0672C CH0672D
470345-648EA 9.8 USD InStock
470345-648 470344-486 470343-694 470343-990
Eisco Support Rings with Bosshead Clamp, Steel
Support Rings
Closed metal support rings with long stems and attached bosshead clamps great for supporting funnels.
  • Closed retort support ring with attached bosshead made from plated mild steel
  • Supports standard or separatory funnels, or use with a triangle to support flat bottomed glassware
  • Distance from rod attachment to center of ring measures 14 cm (5.5"), allowing for vertical alignment across different diameter rings
  • Bosshead fits 8 to 1.2 cm (0.4") diameter rods
  • Stem diameter measures 0.8 cm (0.3")
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty