Would you like to log in again?
Kjeldahl Distillation Apparatus
Supplier: EISCO SCIENTIFIC LLC
CH0885
470344-002EA 174 USD InStock
470344-002
Kjeldahl Distillation Apparatus
Distillation Systems Kjeldahl Distillation Systems
Including a 500 ml Kjeldahl flask, multiple adaptors, a 50 ml dropping funnel, a splash head, and a Liebig condenser, this apparatus with 19/26 joints offers a comprehensive setup for efficient Kjeldahl distillation processes in laboratory applications.
  • Made of high quality, heavy duty 3.3 borosilicate glass
  • 19/26 joint size
  • Liebig condenser and four joint clips
  • Perfect for any classroom or laboratory
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty