Would you like to log in again?
Eisco Two-Hole Rectangular Retort Bases, Steel
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Eisco
CH0656B CH0656C
470344-024EA 21.35 USD InStock
470344-024 470345-122
Eisco Two-Hole Rectangular Retort Bases, Steel
Support Stands
Red steel Retort Bases with two tapped holes support ring stand, titration, or specific gravity setups.
  • Rectangular retort bases made of corrosion resistant, powder coated pressed steel
  • Features two tapped holes - one centered and one at end - for 10×1.5 mm threaded rods (not included)
  • Ideal for supporting standard ring stands or titration and specific gravity setups
  • Sturdy construction provides superb stability for experimental setups
  • Red color
Red-colored Retort Stands are made of corrosion resistant, powder coated pressed steel. They feature two tapped rod holes - one centered and one at a short side end - suitable for rods with 10×1.5 mm metric threads. Provides superb stability for standard ring stands or titration and specific gravity setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty