Would you like to log in again?
LED Ray Box
Supplier: EISCO SCIENTIFIC LLC
PH0602ALED
470344-164EA 47.2 USD InStock
470344-164
LED Ray Box
Educational Materials Physics Educational Materials Optics Learning Activities
Demonstrate the optical principles of lenses and mirrors.
  • Battery operated LED light ray box with seven grooves
  • Ideal for classroom demonstrations in physics, optics, reflection, refraction and more
  • Includes two slit plates and one lens
  • Rugged ABS injection molded enclosure
  • Runs substantially cooler than traditional incandescent lamps
This ray box in an ABS injection molded enclosure, and delivers bright light but runs much cooler than conventional incandescent lamp models. It uses the latest, cool ultra bright LED technology giving superior white light. Supplied complete with lens, two slit plates and its own battery power source.
Ordering information: Batteries are required, but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty