Would you like to log in again?
Eisco Clay Pipe Triangles
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0530B CH0530A CH0530C
470345-128EA 2.29 USD InStock
470345-128 470344-166 470344-366
Eisco Clay Pipe Triangles
Tripods
Braided iron wire wrapped with insulating clay piping on three sides, for supporting hot objects.
  • Holds objects like crucibles on ring stands during or after high heating
  • Made of braided iron wire wrapped with clay piping on three sides
  • Piping measures 11 mm in diameter
Clay Pipe Triangles of varying sizes for supporting very hot items. Made of braided iron wire wrapped with clay piping on its three sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty