Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco® 9-Piece Slotted Mass Set with Hanger, Zinc Casted

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Eisco®
PHZNW1000SET
470344-220EA 44.65 USD InStock
470344-220
Eisco® 9-Piece Slotted Mass Set with Hanger, Zinc Casted
Weights Calibration Weights
Set of 9 slotted masses with a hanger, made of powder-coated, die-cast zinc and stamped for easy identification.

  • Includes 9 individually calibrated 100 g slotted masses and a 100 g mass hanger with hook
  • Total weight of set is 1000 g
  • Made of die-cast zinc and powder-coated to ensure durability and long life
  • Each piece is stamped to identify its mass
  • Good classroom tool for demonstrations on force and balance

Slotted mass set comprised of four individual 100 g masses and one 100 g hanger. Die-cast in zinc and powder-coated to ensure durability and long life. Each piece is stamped to identify its mass value. Masses are slotted allowing individual weights to be removed with ease. Good classroom tool for demonstrations on force and balance.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty