Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Thistle Tube Funnels

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Eisco
CH0489C CH0489A CH0489B
470345-748EA 4.29 USD InStock
470345-748 470343-836 470344-226
Eisco Glass Thistle Tube Funnels
Funnels Filtration Funnels Filter Funnels
Stout-walled Thistle Tube Funnels for the delivery of liquid to an existing system or apparatus.

  • Made of high quality borosilicate 3.3 glass
  • Stout wall design for durability
  • Ideal for filling narrow items like burettes, barometer tubes, and manometers
  • Minimizes the risk of a reaction occurring too rapidly

Thistle Tube funnel for the insertion of liquid to an exact position in an existing system or apparatus. Reservoir is 38 mm in height with a 38 mm Ø and an Int.Ø of 25 mm. The Ext.Ø of the stem is 12 mm.

Eisco borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty