Would you like to log in again?
Eisco Stainless Steel Beaker Tongs
Supplier: EISCO SCIENTIFIC LLC
BKTGSS
470344-228EA 8.09 USD InStock
470344-228
Eisco Stainless Steel Beaker Tongs
Tongs
Corrosion-resistant handle and wide jaw capacity.
  • Extra-long handle made of high-quality, rust- and corrosion-resistant stainless steel
  • Curved jaws with rubber coated tips for added grip strength and safety
  • For use with 50 to 2000 ml capacity beakers
Tongs measure 10" long, 3.5" wide with jaws closed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty