Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco ASTM Glass Graduated Cylinders, Class B

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0341OBL CH0341MBL CH0341IBL CH0341NBL CH0341K CH0341LBL CH0341KBL CH0341JBL
470346-012EA 17.9 USD InStock
470346-012 470344-128 470345-514 470344-240 470344-530 470345-572 470346-106 470345-662
Eisco ASTM Glass Graduated Cylinders, Class B
Cylinders Measuring Cylinders
High precision, durability, and chemical resistance.

  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Conforms with ASTM E1272 Class B tolerance standards
  • Spout for easy pouring
  • Hexagonal base provides maximum stability and prevents rolling when placed on side
  • Easy-to-read graduations and specifications
  • Fitted with a removable, plastic protective collar

Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks. Calibrated to contain.

Eisco labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C

Packaging: One unit per pack.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty