Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Graduated Cylinders with Round Base

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0344LWT CH0344J CH0344N CH0344A10 CH0344P CH0344A10PK12 CH0344OWT CH0344A CH0344O CH0344M CH0344H CH0344L CH0344B CH0344I CH0344K CH0344IWT CH0344C CH0344D CH0344E CH0344HWT CH0344G CH0344A10WT CH0344F CH0344A5
470343-926EA 3.99 USD InStock
470343-926 470345-778 470345-968 470343-768 470343-910 470345-802 470345-948 470345-508 470344-480 470344-062 470344-694 470344-830 470344-246 470344-652 470344-304 470346-034 470345-392 470343-984 470345-186 470346-094 470346-072 470346-090 470345-970 470345-004
Eisco Glass Graduated Cylinders with Round Base
Cylinders Measuring Cylinders
High-quality, round-based borosilicate cylinders with Class A and Class B tolerances conform with DIN ISO standards for accuracy.

  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Available with Class A and Class B tolerances which conform with DIN ISO 4788 standards
  • Spout for easy pouring, and round base
  • Easy-to-read graduations and specifications

Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks. Calibrated to contain.

Eisco Labs Borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty