Would you like to log in again?
Eisco Glass Separatory Funnels with Screw-Type Stopcock
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0484E CH0484C
470344-280EA 23.1 USD InStock
470344-280 470345-882
Eisco Glass Separatory Funnels with Screw-Type Stopcock
Funnels Separating Funnels
Separatory funnels with ground cones, plastic stoppers, and Rotaflow screw-type stopcocks, made of borosilicate 3.3 glass.
  • Made of high quality, heavy-duty borosilicate 3.3 glass - can withstand high temperatures and thermal shocks
  • Used to separate the components of a mixture into two immiscible liquids of different densities
  • Fitted with a screw-type Rotaflow stopcock for precise drop-wise dispensing of the denser separated liquid
  • Features a ground glass cone with and interchangeable plastic stopper
  • Ungraduated
Ungraduated, pear-shaped Separating Funnels are made of high quality borosilicate glass. They feature a ground cone, plastic stopper, and screw-type Rotaflow stopcock. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty