Would you like to log in again?
Distilling Glassware Apparatus
Supplier: EISCO SCIENTIFIC LLC
CH0890
470344-332EA 201.79 USD InStock
470344-332
Distilling Glassware Apparatus
Distillation Components Distillation Sets
With a 500 ml flask, 300 mm condenser, still head, 100 ml separatory funnel, and an adapter, this distilling glassware apparatus with 19/26 joints ensures efficient and precise distillation processes in various laboratory applications.
  • This distilling kit is made of high quality borosilicate glass
  • The kit consists of only the glass ware for distilling experiments
  • No tubing or heat source or stands are included
  • All joint sizes are 19/26
Everything in the picture is included. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Contents:
(1) Distilling 500 ml flask
(1) Three way adapter
(1) Liebig condenser with water inlet and outlet tubes
(1) Vacuum adapter
(1) Separatory funnel 100 ml
(1) Flask stopper
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty