Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Volumetric Pipettes, Class B

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Eisco
CH0558I CH0558E CH0558B CH0558D CH0558G CH0558C CH0558F CH0558A CH0558H
470344-390EA 5.99 USD InStock
470344-390 470344-516 470344-328 470344-678 470345-492 470345-382 470344-342 470345-064 470344-204
Eisco Volumetric Pipettes, Class B
Pipettes Graduated Pipettes
Reusable color-coded Class B pipettes made from high quality borosilicate 3.3 glass.

  • Color coded for easy identification
  • Single blue graduation line
  • Class B, calibrated for delivery
  • Autoclavable
  • Individually wrapped

These Class-B volumetric pipettes are perfect for everyday laboratory use. They are designed for transferring liquid substances from one vessel to another with high accuracy.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty