Would you like to log in again?
Eisco Curved Flask Brush
Supplier: EISCO SCIENTIFIC LLC
CH0216B
470344-406EA 4.69 USD InStock
470344-406
Eisco Curved Flask Brush
Brushes
Designed to scrub the rounded surfaces of flasks, bottles, beakers, cups, jars, dishes, vases and more.
  • Stainless steel, twisted wire handle that is resistant to corrosion
  • Designed for professional laboratories
This semi-stiff, nylon fiber bristle brush is perfect for scrubbing the rounded surfaces of glassware. The stainless steel, twisted handle is semi-flexible, allowing modification to the handle shape as needed, with a looped end for easy hanging. Brush measures 15" in total length. The bristle section measures 6.25" in length and is 1.5" in diameter. Handle measures 8.75" long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty