Would you like to log in again?
Eisco Drechsel Gas Washing Bottles
Supplier: EISCO SCIENTIFIC LLC
CH0196B CH0196A CH0196C
470345-952EA 29.8 USD InStock
470345-952 470345-980 470344-412
Eisco Drechsel Gas Washing Bottles
Bottles Gas Washing Bottles
Remove impurities from gases.
  • Made from high quality borosilicate 3.3 glass
  • Interchangeable ground glass joint allows for easy sealing and exchange
  • Both bottle and joint head are autoclavable
Autoclavable, with superior chemical durability. Low temperature gradient; can withstand high temperatures and thermal shocks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty