Would you like to log in again?
2-Prong Universal Retort Clamp
Supplier: EISCO SCIENTIFIC LLC
CH0688D
470344-468EA 25.55 USD InStock
470344-468
2-Prong Universal Retort Clamp
Clamps General Purpose Clamps
The 2-prong, double adjustable universal clamp offers a non-slip vinyl-coated grip, swivel bosshead, and is made of durable stainless steel.
  • High performance, die casted metal retort clamp, suitable for holding a variety of items. The clamp features two, vinyl coated prongs that are specifically designed for maximum traction and grip. Ideal for use with items that have straight sides
  • Clamp features adjustable thumbscrews on both sides, allowing for a wider range of accommodation
  • Clamp is attached to swivel bosshead that fits rods measuring up to 21 mm in diameter
  • Designed for use in professional laboratories, but is an ideal choice for science classroom
This jumbo size, universal clamp is designed to hold a variety of items. The two, vinyl coated jaws provide a firm grip and are adjustable on both sides with thumbscrews for a wide range of accommodation. Clamp is ideal for use with items that have straight sides, such as burettes, test tubes and flask necks. Accommodates apparatus up to 2.15" (55 mm) wide. Overall length is 8".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty