Would you like to log in again?
Eisco Graduated Micro Centrifuge Tubes
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0284B CH0284A CH0284B CH0284A
470345-986EA 6.6 USD InStock
470345-986 470344-523 470344-522
Eisco Graduated Micro Centrifuge Tubes
Tubes Microcentrifuge Tubes
Pack of 100 graduated, leak-proof micro centrifuge tubes made of polypropylene with raised graduations and snap cap lids.
  • Made of high quality polypropylene
  • Raised graduations
  • Features built in plugs which snap to close the top for a leak proof seal
  • Can withstand high forces of an ultra centrifuge and can be used for transmitting samples for micro analysis
  • Each tube is 1.25" long and 0.3" wide
Pack of 100 Micro Centrifuge Tubes made of high quality polypropylene with raised graduations. Features built in plugs which snap to close the top for a leak proof seal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty