Would you like to log in again?
Micro Glass Distillation Kit
Supplier: EISCO SCIENTIFIC LLC
CH0879
470344-628EA 157.39 USD InStock
470344-628
Micro Glass Distillation Kit
Distillation Components Distillation Sets
Designed for micro-scale distillation, this 9-piece kit includes all the necessary components with 19/26 joints, enabling precise and efficient distillation processes in a compact and convenient setup for laboratory use.
  • Distillation is the process of separating the component or substances from a liquid mixture by selective evaporation and condensation
  • Kit includes 9 pieces : 1 - 50 ml conical flask, 1 - round bottom boiling flask 25 ml, 1 - liebig condenser 100 mm, 1 - still head, 1 - receiver adapter, 1 - funnel 35 mm, 1 - 50 ml beaker, 1 - 10 ml burette ( class A 0.1 ml graduations, ±0.02 tolerance), and 1 - spirit burner
  • Some glass pieces are threaded to make a tight connection with plastic connectors to make a tight fit during operation
  • All pieces are made of high quality borosilicate glass 3.3
  • Clamps, stand, or thermometer are not included with this kit
Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty