Would you like to log in again?
Eisco Plastic Measuring Jugs, Short Form
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0356E CH0356G CH0356F CH0356H CH0356I
470344-656EA 4.49 USD InStock
470344-656 470344-700 470344-916 470343-800 470345-406
Eisco Plastic Measuring Jugs, Short Form
Beakers
Translucent polypropylene measuring jug with short form (Euro design), raised graduations, tapered spout, and handle.
  • Made of premium quality polypropylene plastic
  • Superior chemical durability and heat tolerance (can withstand temperatures up to 135 °C)
  • Autoclave, microwave and dishwasher safe.
  • Excellent resistance to strong acids, alcohols and bases
  • Measuring jug is translucent and features excellent optical clarity with raised graduations and specifications
  • Tapered spout for easy pouring and transferring of liquids
  • Equipped with a handle for easy and convenient lifting
Jug features excellent optical clarity and easy to read, raised graduations and specifications. 50 ml subdivisions. Features a spout for easy pouring, and handle for stable handling and convenient lifting. Microwave and dishwasher safe.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty