Would you like to log in again?
Vacuum Fractional Distillation Kit
Supplier: EISCO SCIENTIFIC LLC
CH1032
470344-702EA 144.39 USD InStock
470344-702
Vacuum Fractional Distillation Kit
Distillation Components Distillation Sets
6-piece kit for vacuum fractional distillation, featuring a 100 ml distillation flask and components with 24/29 joints, made of durable borosilicate glass for reliable performance.
  • 100 ml fractional distillation kit
  • Fractional distillation is the separation of a mixture into individual parts of fractions by heating it up
  • Kit includes 5 pieces: Vigrex column, still head, liebig condenser, receiver adaptor vacuum, 100 ml erlenmeyer flask
  • All joint sizes are 24/29
  • All pieces are made of high quality borosilicate glass
Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty