Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Teclu Burner, Natural Gas

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0096
470344-720EA 61.15 USD InStock
470344-720
Eisco Teclu Burner, Natural Gas
Heaters Gas Burners
Produces very hot flame suitable for sterilization, glasswork, and smoldering.

  • Suitable for use with Natural Gas (methane)
  • Tapered riffled tube connector arm accepts 6 to 8 mm tubing
  • Aluminum mixing tube is mounted on a circular, die casted zinc alloy base
  • Needle valve allows for fine flame adjustment
  • Unique spring operated gas safety lock is incorporated for air mix control

Premium quality Teclu burner that allows you to control both the air mix adjustment and gas flow adjustment. Burner produces a very hot flame and soldering. Burner measures 6.1" tall, and the base measures 3" in Ø.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty