Would you like to log in again?
Glass Tubing Cutter
Supplier: EISCO SCIENTIFIC LLC
CH0759
470344-728EA 14.09 USD InStock
470344-728
Glass Tubing Cutter
Cutters
Featuring an aluminum body and a diamond scribing point, this glass tubing cutter provides precise and clean cuts for various glass tubing applications, ensuring accuracy and efficiency in laboratory work.
  • Diamond scribing point
  • Aluminum body
  • V-groove
  • Suitable for soda and borosilicate tub cutting
Cutter features a diamond scribing point on spring arm and aluminum alloy body with V-groove. Suitable for use with soda and borosilicate glass tubing. Tubing cutter measures 4.25" long, 2.25" tall, and 0.5" wide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty