Would you like to log in again?
Eisco Porcelain Büchner Funnels
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0462G CH0462E CH0462I CH0462F CH0462D CH0462H
470345-192EA 17.6 USD InStock
470345-192 470343-830 470346-062 470345-332 470344-410 470344-762
Eisco Porcelain Büchner Funnels
Funnels Filtration Funnels Büchner Funnels
Büchner funnels with straight sides and fixed perforated plates made of glazed porcelain.
  • For use in filtration (separation of solid matter and liquid)
  • Made of glazed porcelain
  • Features straight side, thick stem, and a fixed perforated plate
  • Filtration paper required, but not included
  • Often used in combination with a Büchner flask, Büchner ring, and sinter seals
Büchner funnels with straight sides and fixed perforated plates are made of glazed porcelain. For use in filtration (separation of solid matter and fluid). Filtration paper required but not included. Moisten filter paper with a liquid to prevent initial leakage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty