Would you like to log in again?
Powder Funnel
Supplier: EISCO SCIENTIFIC LLC
CH0459D
470344-782EA 6.6 USD InStock
470344-782
Powder Funnel
Funnels Powder Funnels
Borosilicate glass powder funnel with an 85 mm diameter and a 60° angle, featuring a plain stem of 24 mm, facilitating smooth and accurate powder transfer in laboratory settings.
  • Powder funnel (60°) with 85 mm diameter and plain stem
  • Features thick, uniform walls that allow for increased durability, ease of use and continual flow of liquid
  • Made of high quality Borosilicate 3.3 glass that is built to last and withstand multiple uses - glass is the material of choice for laboratory applications due to its inertness
In addition to funnel, filter paper is required to correctly separate solids and liquids - can be used with various sizes of paper
Funnel measures 85 mm in diameter - stem has inner diameter measuring 20 mm and outer diameter measuring 24 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty