Would you like to log in again?
Eisco® Aneroid Teaching Barometer
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0248
470344-804EA 36.1 USD InStock
470344-804
Eisco® Aneroid Teaching Barometer
Barometers
Accurately measures barometric pressure and is ideal for classroom use.
  • 13 cm dial numbered in inches and millibar
  • Exposed aneroid shows change in pressure
  • From 28 to 31" of mercury (960 to 1060 millibar)
The EISCO aneroid teaching barometer with pump is a high quality teaching barometer to demonstrate operation. The exposed aneroid shows the change in barometric pressure when you pump the attached bulb.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty