Would you like to log in again?
Eisco Rectangular Retort Bases with Rubber Strips, Steel
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0649C CH0649B CH0649A
470345-624EA 30.2 USD InStock
470345-624 470344-592 470344-818
Eisco Rectangular Retort Bases with Rubber Strips, Steel
Support Stands
Premium Retort Bases with no-slip rubber strips and tapped hole made of powder-coated pressed steel.
  • Rectangular retort bases made of corrosion resistant, powder coated pressed steel
  • Rubber strips on sort ends prevent slipping and protect surfaces
  • Sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Heavy-weighted Retort Stands are made of corrosion resistant, powder coated pressed steel, and feature rubber strips at the short ends prevent slipping and protect surfaces. With a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty