Would you like to log in again?
Plastic Double Burette Clamp
Supplier: EISCO SCIENTIFIC LLC
CH0262B
470344-856EA 8.6 USD InStock
470344-856
Plastic Double Burette Clamp
Clamps General Purpose Clamps
Double burette clamp made of polypropylene plastic, suitable for 10 mm or 12.5 mm rods.
  • Double clamp - Fits 2× rods 10 to 15 mm in diameter
  • Features two tapered wedges and a spring-loaded jack
  • Made of high quality polypropylene
Polypropylene double burette clamps come with easy-to-change wedges that hold rods of 10 mm or 12.5 mm diameter. Rugged yet lightweight, they are equipped with a spring-loaded jack that locks fir mly onto burettes to prevent slip. Non-metal build reduces the chance for accidental impact breaks of glass burettes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty