Would you like to log in again?
Eisco Plastic Volumetric Flasks
Supplier: EISCO SCIENTIFIC LLC
CH0452D CH0452C
470344-142EA 9.5 USD InStock
470344-142 470344-864
Eisco Plastic Volumetric Flasks
Flasks Volumetric Flasks
Polypropylene volumetric flasks with single graduation line and screw caps.
  • Made from high-quality polypropylene
  • Highly chemical resistant, microwavable, and autoclavable at 121 °C
  • Tight-fitting screw cap to prevent leakage of contents
Basic volumetric flasks are made of heavy duty polypropylene plastic with a high heat tolerance, temperature range of −20 to +135 °C, and good to excellent chemical resistance. Autoclavable at 121 °C for easy cleaning and sterilization. Good contact clarity. Fitted with screw cap to prevent leakage. Marked with a single graduation line and provided with a screw cap.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty