Would you like to log in again?
Crucible Tongs with Straight Tips
Supplier: EISCO SCIENTIFIC LLC
CH0732B
470344-964EA 8.29 USD InStock
470344-964
Crucible Tongs with Straight Tips
Tongs
Measuring 12" long, these stainless steel tongs features a bow design, straight serrated tips, and are suitable for various laboratory applications.
  • Crucible tongs made of high-quality, durable stainless steel
  • Straight serrated tips
  • Tongs measure 12" long and open 9" wide
Crucible tongs made of high-quality, durable stainless steel are useful for grasping hot crucibles, flasks, beakers, or dishes. Perfect for any classroom or laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty