Would you like to log in again?
Student Distillation Apparatus Kit
Supplier: EISCO SCIENTIFIC LLC
DIST1SET
470345-020EA 115.59 USD InStock
470345-020
Student Distillation Apparatus Kit
Distillation Components Distillation Sets
Kit includes glassware components, stands, and borosilicate glass for student distillation experiments, providing a comprehensive and reliable setup for educational purposes.
  • Distillation apparatus kit
  • Necessary distillation glassware components and stands
  • Includes retort stand, clamp, tripod burner stand with wire gauze, rubber stoppers, Kjeldahl bulb, Liebig condenser, boiling flask and Erlenmeyer flask
  • Glassware made of high quality, borosilicate glass
  • Great for chemistry classrooms
This distillation apparatus kit includes stands and glassware components, perfect for jump starting your chemistry lab distillation. Set includes retort stand, clamp, tripod stand with wire gauze, rubber stoppers, Kjeldahl bulb, Liebig condenser, 500 ml boiling flask and 500 ml Erlenmeyer flask. Retort stand measures 26" tall, 8" wide and 8" long. Tripod burner stand measures 8" tall, 7" wide and long, and the wire gauze measures 5×5" and features a ceramic center. Liebig condenser measures 18" long. The 500 ml boiling flask, and the 500 ml Erlenmeyer flasks each measure 7" tall, and have bottom diameters of 4". Each flask neck has an inner diameter of 28 mm and an outer diameter measuring 33 mm. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty