Would you like to log in again?
Eisco® 3-Prong Support Clamps, PVC Coated
Supplier: EISCO SCIENTIFIC LLC
Eisco®
CH0684 CH0684A
470344-012EA 12.95 USD InStock
470344-012 470345-024
Eisco® 3-Prong Support Clamps, PVC Coated
Clamps General Purpose Clamps
3-prong retort support clamps with angled, PVC coated jaws that open to hold items up to 90 mm in diameter.
  • 3-Prong Support Clamp made of diecast alloy
  • Holds items up to 90 mm in diameter
  • PVC coated jaws protect glass from scratches
  • Angled jaw design ensures a first grip at all angles of opening
  • Overall length is 25 cm
The 3-Prong Support Clamp is ideal for clamping glassware of various sizes onto retort rods. The PVC coating on the jaws protects vessels from being scratched. Clamp has a 5.65" long rod which is 8mm in diameter and will fit into most bossheads. Adjustable jaw capacity is 0 to 90 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty