Would you like to log in again?
Eisco Glass Prism and Lens Set, 7-Pieces
Supplier: EISCO SCIENTIFIC LLC
Eisco
PH0572AG
470345-096EA 55.15 USD InStock
470345-096
Eisco Glass Prism and Lens Set, 7-Pieces
Educational Materials Physics Educational Materials Optics Learning Activities
A variety of lenses and prism block shapes, packed in a wood storage case.
  • Ideal for a variety of experiments in optics, light refraction, spectrometry and wavelengths
  • Made of highly-polished glass
  • Comes in a sturdy, polished wooden storage box with custom foam inserts
Set of 7 optics elements in a wooden storage box. Each of the elements is made of durable, highly polished glass and measures 0.5" (13 mm) thick. Excellent for physics classrooms, as well as photography lighting and distortion effects.
Delivery information: Kit includes (1) equilateral trianglular prism, 3"; (1) triangular prism, 2×1×0.5"; (1) triangular prism, 2×1×0.75"; (1 ) trapezoid, 1.3" and 1.6" legs; (1) semi-circle, Ø 3.5"; (1) double convex prism, 3.5" with a 3.75" inner radius; (1) double convex prism, 3.5" with a 3.75" outer radius.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty