Would you like to log in again?
Eisco® Slotted Mass Sets with Hangers, Brass
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Eisco®
PH0258A PH0258B PH0258D PH0258C PH0258F
470345-140EA 32.8 USD InStock
470345-140 470345-628 470344-022 470344-796 470344-960
Eisco® Slotted Mass Sets with Hangers, Brass
Weights Calibration Weights
Slotted mass sets with hangers made of brass and engraved for identification.
  • Includes individually calibrated slotted masses and a mass hanger with hook
  • Total weight of set includes hanger
  • Made of high-quality brass with a fine finish
  • Each piece is engraved to identify its mass
  • An essential piece of equipment for many weight-bearing science experiments spanning elementary to university learning
Slotted mass sets comprised of individual masses and one hanger. Made of high-quality brass with a fine finish. Each piece is engraved to identify its mass value. Masses are slotted allowing individual weights to be removed with ease. An essential piece of equipment for many weight-bearing science experiments spanning elementary to university learning.
Caution: Warning: cancer and reproductive harm (www.p65warnings.ca.gov).
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty