Would you like to log in again?
Eisco Rectangular Retort Bases, Steel
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0652D CH0652B CH0652C CH0652A
470345-608EA 23.5 USD InStock
470345-608 470345-144 470344-886 470343-794
Eisco Rectangular Retort Bases, Steel
Support Stands
Economical Retort Bases with tapped rod hole made of powder-coated pressed steel.
  • Rectangular retort bases made of corrosion resistant, powder coated pressed steel
  • Sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Retort Stands are made of corrosion resistant, powder coated pressed steel, and feature a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty