Would you like to log in again?
Eisco Evaporating Flasks
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0435F CH0435C CH0435B CH0435A CH0435E CH0435D CH0435G
470343-690EA 21.8 USD InStock
470343-690 470344-070 470343-604 470345-150 470345-262 470344-674 470344-872
Eisco Evaporating Flasks
Flasks Rotary Evaporator Flasks
Evaporating flasks with ground sockets made of borosilicate 3.3 glass.
  • Withstands high temperatures and thermal shocks
  • Ungraduated, with round bottom
  • Features ground socket
  • Used with rotary evaporators and vacuum applications that vaporize solvents
Evaporating flasks for use with rotary evaporators and vacuum applications that vaporize solvents. Made of durable, high quality, heavy duty 3.3 borosilicate glass with the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty