Would you like to log in again?
Sampling Tongs
Supplier: EISCO SCIENTIFIC LLC
CH0736
470345-158EA 4.9 USD InStock
470345-158
Sampling Tongs
Tongs
These nickel-plated tongs are ideal for sampling purposes, providing a reliable and precise grip for various materials in laboratory and industrial environments.
  • 8.25" long sampling spoon/tongs
  • 1" spoon splits in half vertically; useful for collecting hot samples of thick materials
  • Designed to handle mercury safely
  • Scalloped spoon design; half scallop splits into a quarter per side
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty