Would you like to log in again?
7-Piece Filter Adapter Tapered Cones Set
Supplier: EISCO SCIENTIFIC LLC
CH0421
470345-184EA 17.1 USD InStock
470345-184
7-Piece Filter Adapter Tapered Cones Set
Funnels Filtration Funnels
Set of 7 neoprene rubber tapered cones specifically designed for Buchner funnels, providing a secure and efficient filtration system, available in multiple sizes for versatile laboratory applications.
  • Size 1 to 7 tapered, blue-colored filter adapter cones
  • Designed for use with Buchner funnels in filtration assemblies
  • Made of quality, flexible neoprene rubber - Good chemical stability and can withstand a wide temperature range - Autoclavable
  • Can be nested within different size cones for convenient storage
  • Cones vary in size - Height: 20 to 43 mm, Top: 22 to 81 mm, Bottom: 11 to 57 mm
Set of 7 tapered filter adapter cones. These adapter cones are made for use with Buchner Funnels in filtration assemblies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty