Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Bunsen Burner

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0088C CH0088D
470344-776EA 27.6 USD InStock
470344-776 470345-200
Eisco Bunsen Burner
Heaters Gas Burners
Economical option for general laboratory applications.

  • 11 mm burner tube does not have a flame stabilizer
  • Serrated inlet arm that accepts 6 to 8 mm tubing
  • Mounted on a stable, circular base

Economical, laboratory quality gas burner ideal for general lab applications. Burner comes equipped with a serrated inlet arm that measures 8 mm in Ø and 1" in length. No flame stabilizer or gas adjustment. Burner tube Ø is 13 mm.

Overall size: 5.7tall×3" Ø base.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty