Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Tirrill Burner

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0095B CH0095A
470345-230EA 52.75 USD InStock
470345-230 470345-680
Eisco Tirrill Burner
Heaters Gas Burners
Designed to provide the most precise control of flame.

  • Both air and gas supply can be adjusted, allowing for separate control of flame height and temperature
  • Gas control provided by a thumb knob on the bottom of the burner
  • Adjustable air ports control air supply
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Highly stable octagonal base

An additional level of control is gained when using a Tirrill Burner instead of a standard burner. Burner measures 6.25" tall, and the base measures 3.25" wide.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty